MAN238929472 Manchester Organics

2,6-Dichloro-4-methoxypyridine

  • MDL number MFCD00955755
  • Molecular Weight : 178.016

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V19414
Availability details for
Quantity
  • Availability
  • Add to Cart