MAN238929482 Manchester Organics

1-[4-(2-Aminophenyl)piperazin-1-yl]ethanone

  • MDL number MFCD03848245
  • Molecular Weight : 219.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E19494

Safety Information

MSDS https://www.manchesterorganics.com/E19494
Availability details for
Quantity
  • Availability
  • Add to Cart