MAN238929485 Manchester Organics

3-Bromo-2-fluoro-5-methylpyridine

  • MDL number MFCD03095305
  • Cas Number : 17282-01-8
  • Molecular Weight : 190.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-2-fluoro-5-methylpyridine
Inchi Code 1S/C6H5BrFN/c1-4-2-5(7)6(8)9-3-4/h2-3H,1H3
Inchi Key FWKQBHMQYXTOTD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/A19419
Availability details for
Quantity
  • Availability
  • Add to Cart