MAN238929500 Manchester Organics

Amino-naphthalen-2-yl-acetic acid

  • MDL number MFCD02662462
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y18897
Availability details for
Quantity
  • Availability
  • Add to Cart