MAN238929506 Manchester Organics

3,5-Dichloro-4-fluoropyridine

  • MDL number MFCD07368620
  • Molecular Weight : 165.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B19468

Safety Information

MSDS https://www.manchesterorganics.com/B19468
Availability details for
Quantity
  • Availability
  • Add to Cart