MAN238929562 Manchester Organics

3,4-Dichloro-2,5,6-trifluoropyridine

  • MDL number MFCD12024847
  • Molecular Weight : 201.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H19545

Safety Information

MSDS https://www.manchesterorganics.com/H19545
Availability details for
Quantity
  • Availability
  • Add to Cart