MAN238929574 Manchester Organics

4-Bromo-2-(trifluoromethoxy)benzyl alcohol

  • MDL number MFCD12546981
  • Molecular Weight : 271.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J19617

Safety Information

MSDS https://www.manchesterorganics.com/J19617
Availability details for
Quantity
  • Availability
  • Add to Cart