MAN238929612 Manchester Organics

Ethyl ethoxymethylene-3-oxo-4,4,4-trifluoro-butyrate

  • MDL number MFCD00052716
  • Cas Number : 571-55-1
  • Molecular Weight : 240.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/A10995
Availability details for
Quantity
  • Availability
  • Add to Cart