MAN238929617 Manchester Organics

D,L-Anatabine

  • MDL number MFCD00078278
  • Molecular Weight : 160.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D10377

Safety Information

MSDS https://www.manchesterorganics.com/D10377
Availability details for
Quantity
  • Availability
  • Add to Cart