MAN238929621 Manchester Organics

1H-Pyrrolo[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD06659752
  • Molecular Weight : 162.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K19570

Safety Information

MSDS https://www.manchesterorganics.com/K19570
Availability details for
Quantity
  • Availability
  • Add to Cart