MAN238929626 Manchester Organics

2-Amino-3,5,6-trifluoropyridine

  • MDL number MFCD03001164
  • Cas Number : 3534-50-7
  • Molecular Weight : 148.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/P19457
Availability details for
Quantity
  • Availability
  • Add to Cart