MAN238929647 Manchester Organics

4-Methoxy-2-(trifluoromethoxy)benzoic acid

  • MDL number MFCD06660285
  • Molecular Weight : 236.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D19589

Safety Information

MSDS https://www.manchesterorganics.com/D19589
Availability details for
Quantity
  • Availability
  • Add to Cart