MAN238929649 Manchester Organics

5-Bromo-2,4-difluoropyrimidine

  • MDL number MFCD07787386
  • Molecular Weight : 194.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U19461
Availability details for
Quantity
  • Availability
  • Add to Cart