MAN238929650 Manchester Organics

4-Bromo-2-(trifluoromethoxy)benzoic acid

  • MDL number MFCD12546979
  • Molecular Weight : 285.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/G19521
Availability details for
Quantity
  • Availability
  • Add to Cart