MAN238929675 Manchester Organics

6-Ethyl-2-oxo-1,2-dihydro-pyridine-4-carboxylic acid

  • MDL number MFCD11226364
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L19642

Safety Information

MSDS https://www.manchesterorganics.com/L19642
Availability details for
Quantity
  • Availability
  • Add to Cart