MAN238929686 Manchester Organics

Methyl-piperidine-2-carboxylate

  • MDL number MFCD07364347
  • Cas Number : 41994-45-0
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-piperidinecarboxylate
Inchi Code 1S/C7H13NO2/c1-10-7(9)6-4-2-3-5-8-6/h6,8H,2-5H2,1H3
Inchi Key CXQTTWVBUDFUNO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/C19563
Availability details for
Quantity
  • Availability
  • Add to Cart