MAN238929698 Manchester Organics

5-Chloro-2-(trifluoromethyl)benzaldehyde

  • MDL number MFCD03412204
  • Molecular Weight : 208.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P19741

Safety Information

MSDS https://www.manchesterorganics.com/P19741
Availability details for
Quantity
  • Availability
  • Add to Cart