MAN238929707 Manchester Organics

1-Methylnaphthalene-1-methylamine

  • MDL number MFCD00144934
  • Cas Number : 14489-75-9
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/W19580
Availability details for
Quantity
  • Availability
  • Add to Cart