MAN238929710 Manchester Organics

1-Methyl-1H-indole-5-carbonitrile

  • MDL number MFCD09025840
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z20057

Safety Information

MSDS https://www.manchesterorganics.com/Z20057
Availability details for
Quantity
  • Availability
  • Add to Cart