MAN238929711 Manchester Organics

5-Bromo-4-fluoro-2-iodotoluene

  • MDL number MFCD09800686
  • Molecular Weight : 314.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y19914

Safety Information

MSDS https://www.manchesterorganics.com/Y19914
Availability details for
Quantity
  • Availability
  • Add to Cart