MAN238929717 Manchester Organics

5-Bromo-2-hydroxyisonicotinic acid

  • MDL number MFCD08689503
  • Molecular Weight : 218.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C19634

Safety Information

MSDS https://www.manchesterorganics.com/C19634
Availability details for
Quantity
  • Availability
  • Add to Cart