MAN238929726 Manchester Organics

2-Furan-2-ylpyrrolidine hydrochloride

  • MDL number MFCD06660476
  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L19690

Safety Information

MSDS https://www.manchesterorganics.com/L19690
Availability details for
Quantity
  • Availability
  • Add to Cart