MAN238929728 Manchester Organics

2-Methoxypyridine-4-boronic acid

  • MDL number MFCD07368877
  • Cas Number : 762262-09-9
  • Molecular Weight : 152.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/N19598
Availability details for
Quantity
  • Availability
  • Add to Cart