MAN238929730 Manchester Organics

4-Bromo-2-fluoro-5-methylbenzonitrile

  • MDL number MFCD07782073
  • Cas Number : 916792-13-7
  • Molecular Weight : 214.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/T19602
Availability details for
Quantity
  • Availability
  • Add to Cart