MAN238929736 Manchester Organics

2-Fluoro-3-nitro-5-bromopyridine

  • MDL number MFCD07375020
  • Molecular Weight : 220.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X19606
Availability details for
Quantity
  • Availability
  • Add to Cart