MAN238929805 Manchester Organics

5-Bromo-3-piperidin-1-yl-pyrazin-2-ylamine

  • MDL number MFCD08460061
  • Molecular Weight : 257.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D19754

Safety Information

MSDS https://www.manchesterorganics.com/D19754
Availability details for
Quantity
  • Availability
  • Add to Cart