MAN238929872 Manchester Organics

3,5-Dibromo-pyrazine-2-carboxylic acid methyl ester

  • MDL number MFCD11226366
  • Molecular Weight : 295.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P19716

Safety Information

MSDS https://www.manchesterorganics.com/P19716
Availability details for
Quantity
  • Availability
  • Add to Cart