MAN238929918 Manchester Organics

5-Benzylsulfanyl-[1,3,4]thiadiazol-2-ylamine

  • MDL number MFCD00184237
  • Molecular Weight : 223.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L19855

Safety Information

MSDS https://www.manchesterorganics.com/L19855
Availability details for
Quantity
  • Availability
  • Add to Cart