MAN238929930 Manchester Organics

2,4-Dimethylnicotinic acid

  • MDL number MFCD03265250
  • Cas Number : 55314-30-2
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/T19815
Availability details for
Quantity
  • Availability
  • Add to Cart