MAN238929938 Manchester Organics

7-Bromobenzo[b]thiophene

  • MDL number MFCD06657729
  • Cas Number : 1423-61-6
  • Molecular Weight : 213.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/B19823
Availability details for
Quantity
  • Availability
  • Add to Cart