MAN238929950 Manchester Organics

4-Oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD01794098
  • Molecular Weight : 190.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N19928

Safety Information

MSDS https://www.manchesterorganics.com/N19928
Availability details for
Quantity
  • Availability
  • Add to Cart