MAN238930000 Manchester Organics

1,3-Thiazole-2-carboxamide

  • MDL number MFCD08361702
  • Molecular Weight : 128.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N19882
Availability details for
Quantity
  • Availability
  • Add to Cart