MAN238930013 Manchester Organics

(2S,4R)-1-(Tert-butoxycarbonyl)-4-hydroxypiperidine-2-carboxylic acid

  • MDL number MFCD04112667
  • Molecular Weight : 245.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z20389

Safety Information

MSDS https://www.manchesterorganics.com/Z20389
Availability details for
Quantity
  • Availability
  • Add to Cart