MAN238930019 Manchester Organics

4-(4-Amino-3-nitrophenyl)-2-(tert-butoxycarbonylamino) butanoic acid

  • MDL number MFCD12404979
  • Molecular Weight : 339.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T20170

Safety Information

MSDS https://www.manchesterorganics.com/T20170
Availability details for
Quantity
  • Availability
  • Add to Cart