MAN238930035 Manchester Organics

(1-Cyano-cyclopentyl)-carbamic acidtert-butyl ester

  • MDL number MFCD08277592
  • Molecular Weight : 210.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J20043

Safety Information

MSDS https://www.manchesterorganics.com/J20043
Availability details for
Quantity
  • Availability
  • Add to Cart