MAN238930050 Manchester Organics

3-Bromo-7-methyl-imidazo[1,2-a]pyridine

  • MDL number MFCD08056208
  • Molecular Weight : 211.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V20337

Safety Information

MSDS https://www.manchesterorganics.com/V20337
Availability details for
Quantity
  • Availability
  • Add to Cart