MAN238930056 Manchester Organics

6-(4-Methoxy-phenyl)-3-pyridin-3-yl-pyrazolo[1,5-a]pyrimidine

  • MDL number MFCD08056197
  • Molecular Weight : 302.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J20091

Safety Information

MSDS https://www.manchesterorganics.com/J20091
Availability details for
Quantity
  • Availability
  • Add to Cart