MAN238930090 Manchester Organics

1,3-Thiazole-2-thiocarboxamide

  • MDL number MFCD06738992
  • Cas Number : 60758-41-0
  • Molecular Weight : 144.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/M19881
Availability details for
Quantity
  • Availability
  • Add to Cart