MAN238930092 Manchester Organics

5-Acetamido-1-indanone

  • MDL number MFCD00099465
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R20049

Safety Information

MSDS https://www.manchesterorganics.com/R20049
Availability details for
Quantity
  • Availability
  • Add to Cart