MAN238930103 Manchester Organics

3-Benzylpiperidine-3-carboxylic acid ethyl ester hydrochloride

  • MDL number MFCD03840052
  • Cas Number : 170842-81-6
  • Molecular Weight : 283.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/J11003
Availability details for
Quantity
  • Availability
  • Add to Cart