MAN238930121 Manchester Organics

2,6-Difluoropyridine-4-carboxylic acid

  • MDL number MFCD09864665
  • Molecular Weight : 159.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M19430

Safety Information

MSDS https://www.manchesterorganics.com/M19430
Availability details for
Quantity
  • Availability
  • Add to Cart