MAN238930123 Manchester Organics

2-Amino-4,5-difluorobenzoic acid methyl ester

  • MDL number MFCD09264511
  • Cas Number : 207346-42-7
  • Molecular Weight : 187.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-4,5-difluorobenzoate
Inchi Code 1S/C8H7F2NO2/c1-13-8(12)4-2-5(9)6(10)3-7(4)11/h2-3H,11H2,1H3
Inchi Key VIAQNTRKUPBQKR-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin GB
Product Link https://www.manchesterorganics.com/P20048
Availability details for
Quantity
  • Availability
  • Add to Cart