MAN238930135 Manchester Organics

Methyl 4-(4-amino-3-nitrophenyl)-2-(tert-butoxycarbonylamino)butanoate

  • MDL number MFCD12404978
  • Molecular Weight : 353.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S20121

Safety Information

MSDS https://www.manchesterorganics.com/S20121
Availability details for
Quantity
  • Availability
  • Add to Cart