MAN238930136 Manchester Organics

1-Boc-4-(6-Aminopyridin-3-yl)piperazine

  • MDL number MFCD11594962
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U20217

Safety Information

MSDS https://www.manchesterorganics.com/U20217
Availability details for
Quantity
  • Availability
  • Add to Cart