MAN238930150 Manchester Organics

Methyl-3-ketocholate

  • MDL number MFCD00200688
  • Molecular Weight : 420.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M20069

Safety Information

MSDS https://www.manchesterorganics.com/M20069
Availability details for
Quantity
  • Availability
  • Add to Cart