MAN238930160 Manchester Organics

1-tert-Butoxycarbonylamino-cycloheptanecarboxylic acid

  • MDL number MFCD03426130
  • Molecular Weight : 257.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B20011

Safety Information

MSDS https://www.manchesterorganics.com/B20011
Availability details for
Quantity
  • Availability
  • Add to Cart