MAN238930201 Manchester Organics

3-Hydroxy-2-(4-methoxy-phenyl)-propenal

  • MDL number MFCD00832061
  • Molecular Weight : 178.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y20505

Safety Information

MSDS https://www.manchesterorganics.com/Y20505
Availability details for
Quantity
  • Availability
  • Add to Cart