MAN238930212 Manchester Organics

6-Chloro-3H-quinazolin-4-one

  • MDL number MFCD00182168
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F20134

Safety Information

MSDS https://www.manchesterorganics.com/F20134
Availability details for
Quantity
  • Availability
  • Add to Cart