MAN238930228 Manchester Organics

2-Propyl-1,3-thiazole

  • MDL number MFCD00053154
  • Molecular Weight : 127.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y20079
Availability details for
Quantity
  • Availability
  • Add to Cart