MAN238930235 Manchester Organics

6-Quinolinecarbonitrile

  • MDL number MFCD09999543
  • Molecular Weight : 154.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F20159

Safety Information

MSDS https://www.manchesterorganics.com/F20159
Availability details for
Quantity
  • Availability
  • Add to Cart